missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ tert-Butyl (S)-(+)-3-hydroxybutyrate, 98%

$206.65 - $829.66

Chemical Identifiers

CAS 82578-45-8
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.213
MDL Number MFCD00040559
InChI Key PKPVFILAHLKSNJ-LURJTMIESA-N
Synonym s-tert-butyl 3-hydroxybutanoate, s-3-hydroxy-butyric acid tert-butyl ester, tert-butyl 3s-3-hydroxybutanoate, +-tert-butyl s-3-hydroxybutyrate, s-3-hydroxybutyric acid tert-butyl ester, butanoic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 3s, s-t-butyl-3-hydroxybutanoate, s-tert-butyl3-hydroxybutanoate, tert-butyl s-3-hydroxybutyrate, +-tert-butyl s-3-hydroxybutyrate
PubChem CID 10154184
IUPAC Name tert-butyl (3S)-3-hydroxybutanoate
SMILES CC(CC(=O)OC(C)(C)C)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6001303 View Documents Alfa Aesar™
H6001303
1g Each for $206.65
 
AAH6001306 View Documents Alfa Aesar™
H6001306
5g Each for $829.66
 
Specifications

Chemical Identifiers

82578-45-8
160.213
PKPVFILAHLKSNJ-LURJTMIESA-N
10154184
CC(CC(=O)OC(C)(C)C)O
C8H16O3
MFCD00040559
s-tert-butyl 3-hydroxybutanoate, s-3-hydroxy-butyric acid tert-butyl ester, tert-butyl 3s-3-hydroxybutanoate, +-tert-butyl s-3-hydroxybutyrate, s-3-hydroxybutyric acid tert-butyl ester, butanoic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 3s, s-t-butyl-3-hydroxybutanoate, s-tert-butyl3-hydroxybutanoate, tert-butyl s-3-hydroxybutyrate, +-tert-butyl s-3-hydroxybutyrate
tert-butyl (3S)-3-hydroxybutanoate

Specifications

1g
C8H16O3
s-tert-butyl 3-hydroxybutanoate, s-3-hydroxy-butyric acid tert-butyl ester, tert-butyl 3s-3-hydroxybutanoate, +-tert-butyl s-3-hydroxybutyrate, s-3-hydroxybutyric acid tert-butyl ester, butanoic acid, 3-hydroxy-, 1,1-dimethylethyl ester, 3s, s-t-butyl-3-hydroxybutanoate, s-tert-butyl3-hydroxybutanoate, tert-butyl s-3-hydroxybutyrate, +-tert-butyl s-3-hydroxybutyrate
CC(CC(=O)OC(C)(C)C)O
160.213
160.21
tert-Butyl (S)-(+)-3-hydroxybutyrate
82578-45-8
MFCD00040559
PKPVFILAHLKSNJ-LURJTMIESA-N
tert-butyl (3S)-3-hydroxybutanoate
10154184
98%