missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl (S)-2-aminobutyrate hydrochloride, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6528403

 View more versions of this product

Catalog No. AAH6528403



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

tert-Butyl (S)-2-aminobutyrate hydrochloride
C8H18ClNO2
1 g
h-abu-otbu.hcl, s-tert-butyl 2-aminobutanoate hydrochloride, h-2-abu-otbu.hcl, h-2-abu-otbu hcl, tert-butyl 2s-2-aminobutanoate hydrochloride, s-2-aminobutyric acid t-butyl ester hydrochloride, butanoic acid, 2-amino-, 1,1-dimethylethyl ester, hydrochloride, s, 2s-2-aminobutyric acid tert-butyl ester hydrochloride, h-abu-otbu hcl, h-2-abu-otbu hydrochloride
CCC(C(=O)OC(C)(C)C)N.Cl
195.687
195.69
Solid
53956-05-1
MFCD00237697
6935251
OEPKETQBSXWOBJ-RGMNGODLSA-N
tert-butyl (2S)-2-aminobutanoate;hydrochloride
53628861
95%

Chemical Identifiers

53956-05-1
195.687
OEPKETQBSXWOBJ-RGMNGODLSA-N
53628861
CCC(C(=O)OC(C)(C)C)N.Cl
C8H18ClNO2
MFCD00237697
h-abu-otbu.hcl, s-tert-butyl 2-aminobutanoate hydrochloride, h-2-abu-otbu.hcl, h-2-abu-otbu hcl, tert-butyl 2s-2-aminobutanoate hydrochloride, s-2-aminobutyric acid t-butyl ester hydrochloride, butanoic acid, 2-amino-, 1,1-dimethylethyl ester, hydrochloride, s, 2s-2-aminobutyric acid tert-butyl ester hydrochloride, h-abu-otbu hcl, h-2-abu-otbu hydrochloride
tert-butyl (2S)-2-aminobutanoate;hydrochloride
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions