Please login to your online account to display your discounted pricing

Alfa Aesar™ tert-Butyl (S)-2-aminobutyrate hydrochloride, 95%

Chemical Identifiers

Chemical Name or Material tert-Butyl (S)-2-aminobutyrate hydrochloride
CAS 53956-05-1
Formula Weight 195.69
MDL Number MFCD00237697
Beilstein 6935251
Synonym H-2-Abu-OtBu.HCl; (S)-2-Aminobutyric acid tert-butyl ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6528403

  SDS
alfa aesar™
H6528403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6528406

  SDS
alfa aesar™
H6528406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6528414

  SDS
alfa aesar™
H6528414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 195.69
CAS 53956-05-1
MDL Number MFCD00237697
Physical Form Solid
Assay 95%
Beilstein 6935251
Quantity 1g
Storage Note 1 2° to 8°C
Synonym H-2-Abu-OtBu.HCl; (S)-2-Aminobutyric acid tert-butyl ester hydrochloride
Chemical Name or Material tert-Butyl (S)-2-aminobutyrate hydrochloride
Safety and Handling

2° to 8°C