missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl (S)-2-(3-amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetate, 98%, Alfa Aesar™

Manufacturer:  Alfa Aesar H6165522

 View more versions of this product

Catalog No. AAH6165522



Specifications

Specifications

tert-Butyl (S)-2-(3-amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetate
100g
C16H22N2O3
1,1-dimethylethyl 3s-3-amino-2-oxo-2,3,4,5-tetrahydro-1h-1-benzazepin-1-yl acetate
CC(C)(C)OC(=O)CN1C2=CC=CC=C2CCC(C1=O)N
290.363
290.36
98%
White
109010-60-8
MFCD03426719
QTEDVVHLTMELTB-LBPRGKRZSA-N
tert-butyl 2-[(3S)-3-amino-2-oxo-4,5-dihydro-3H-1-benzazepin-1-yl]acetate
6350624
Powder

Chemical Identifiers

109010-60-8
290.363
QTEDVVHLTMELTB-LBPRGKRZSA-N
6350624
CC(C)(C)OC(=O)CN1C2=CC=CC=C2CCC(C1=O)N
C16H22N2O3
MFCD03426719
1,1-dimethylethyl 3s-3-amino-2-oxo-2,3,4,5-tetrahydro-1h-1-benzazepin-1-yl acetate
tert-butyl 2-[(3S)-3-amino-2-oxo-4,5-dihydro-3H-1-benzazepin-1-yl]acetate
Documents