Please login to your online account to display your discounted pricing

Alfa Aesar™ tert-Butyl (S)-2-(3-amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetate, 98%

Chemical Identifiers

Chemical Name or Material tert-Butyl (S)-2-(3-amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetate
CAS 109010-60-8
Formula Weight 290.36
MDL Number MFCD03426719
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6165506

 
alfa aesar™
H6165506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6165514

 
alfa aesar™
H6165514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6165522

 
alfa aesar™
H6165522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 290.36
Chemical Name or Material tert-Butyl (S)-2-(3-amino-2-oxo-2,3,4,5-tetrahydro-1H-benzo[b]azepin-1-yl)acetate
Physical Form Powder
Assay 98%
MDL Number MFCD03426719
CAS 109010-60-8
Color White
Quantity 5g