missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl (R)-(+)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate, 95%, ACROS Organics™

$314.24 - $314.24

Chemical Identifiers

CAS 95715-87-0
Assay Percent Range 95%
Molecular Formula C11H19NO4
Formula Weight 229.28
MDL Number MFCD00674064
Chemical Name or Material tert-Butyl (R)-(+)-4-formyl-2, 2-dimethyl-3-oxazolidinecarboxylate
Assay ≥94%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC370890010 SDS Thermo Fisher Scientific
AC370890010
Glass bottle 1g Each for $314.24
Specifications Description & Specifications

Chemical Identifiers

95715-87-0
C11H19NO4
MFCD00674064
≥94%
95%
229.28
tert-Butyl (R)-(+)-4-formyl-2, 2-dimethyl-3-oxazolidinecarboxylate

Specifications

95715-87-0
Authentic
+85° to +95° (20°C, 589nm) (c=1, CHCl3)
Clear colorless to pale yellow
1g
C11H19NO4
MFCD00674064
tert-Butyl (R)-(+)-4-formyl-2, 2-dimethyl-3-oxazolidinecarboxylate
1.06g/mL
Glass bottle
1.4450 to 1.4490 (20°C, 589nm)
Oil
95%
229.28
15, 4404
≥94%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.