missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ tert-Butyl propargyl ether, 98%

Manufacturer:  Alfa Aesar™ H5348014

 View more versions of this product

Catalog No. AAH5348014



Specifications

Specifications

tert-Butyl propargyl ether
20°C (68°F)
119°C
16314-18-4
HC≡CCH2OC(CH3)3
UN1993
QIGSPXPODKLGRK-UHFFFAOYSA-N
2-methyl-2-prop-2-ynoxypropane
7172302
98%
0.835
1.418
25g
C7H12O
MFCD07368651
1-propyne, 3-1,1-dimethylethoxy
CC(C)(C)OCC#C
112.172
112.17

Chemical Identifiers

16314-18-4
112.172
QIGSPXPODKLGRK-UHFFFAOYSA-N
7172302
CC(C)(C)OCC#C
C7H12O
MFCD07368651
1-propyne, 3-1,1-dimethylethoxy
2-methyl-2-prop-2-ynoxypropane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents