missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ tert-Butyl propargyl ether, 98%

Manufacturer:  Alfa Aesar™ H5348006

 View more versions of this product

Catalog No. AAH5348006



Specifications

Specifications

tert-Butyl propargyl ether
119°C
16314-18-4
HC≡CCH2OC(CH3)3
UN1993
QIGSPXPODKLGRK-UHFFFAOYSA-N
2-methyl-2-prop-2-ynoxypropane
7172302
98%
20°C (68°F)
5g
C7H12O
MFCD07368651
1-propyne, 3-1,1-dimethylethoxy
CC(C)(C)OCC#C
112.172
112.17

Chemical Identifiers

16314-18-4
112.172
QIGSPXPODKLGRK-UHFFFAOYSA-N
7172302
CC(C)(C)OCC#C
C7H12O
MFCD07368651
1-propyne, 3-1,1-dimethylethoxy
2-methyl-2-prop-2-ynoxypropane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Documents