Please login to your online account to display your discounted pricing

Alfa Aesar™ tert-Butyl phenyl ether, 98%

$210.88 - $1,630.09

Chemical Identifiers

Chemical Name or Material tert-Butyl phenyl ether
CAS 6669-13-2
Molecular Formula C10H14O
Formula Weight 150.22
MDL Number MFCD00049018
Synonym tert-Butoxybenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2768903

 
alfa aesar™
H2768903
1g Each for $531.05

AAH27689MD

 
alfa aesar™
H27689MD
250mg Each for $210.88

AAH2768906

 
alfa aesar™
H2768906
5g Each for $1,630.09
Description & Specifications

Specifications

Formula Weight 150.22
Chemical Name or Material tert-Butyl phenyl ether
Assay 98%
Molecular Formula C10H14O
MDL Number MFCD00049018
CAS 6669-13-2
Synonym tert-Butoxybenzene
Boiling Point 184°C
Quantity 1g