missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl phenyl carbonate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC154390250

 View more versions of this product

Catalog No. AC154390250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

tert-Butyl phenyl carbonate
101°C
Glass bottle
1.05
Colorless
6627-89-0
C11H14O3
05,86
UXWVQHXKKOGTSY-UHFFFAOYSA-N
tert-butyl phenyl carbonate
81113
Liquid or Low Melting Solid
1.0500g/mL
Authentic
1.4805
83.0°C (0.6 mmHg)
25g
97.5% min. (GC)
C6H5OCO2C(CH3)3
2-methyl-2-propanyl phenyl carbonate
CC(C)(C)OC(=O)OC1=CC=CC=C1
194.23
194.23
98%

Chemical Identifiers

6627-89-0
194.23
2-methyl-2-propanyl phenyl carbonate
tert-butyl phenyl carbonate
C11H14O3
UXWVQHXKKOGTSY-UHFFFAOYSA-N
81113
CC(C)(C)OC(=O)OC1=CC=CC=C1
Safety and Handling

Safety and Handling

EINECSNumber : 229-601-8

SDS Documents