Please login to your online account to display your discounted pricing

tert-Butyl phenyl carbonate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material tert-Butyl phenyl carbonate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H14O3
Linear Formula C6H5OCO2C(CH3)3
Formula Weight 194.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC154391000

  SDS
thermo fisher scientific
AC154391000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC154390250

  SDS
thermo fisher scientific
AC154390250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 194.23
Physical Form Liquid or Low Melting Solid
Quantity 100g
Color Colorless
Density 1.05g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
CAS 6627-89-0
Chemical Name or Material tert-Butyl phenyl carbonate
Molecular Formula C11H14O3
Linear Formula C6H5OCO2C(CH3)3