Learn More
tert-Butyl peroxybenzoate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 170142500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Radical initiatorsSpecifications
tert-Butyl peroxybenzoate | |
614-45-9 | |
Yellow | |
75.0°C to 76.0°C (0.2 mmHg) | |
Authentic | |
Plastic bottle | |
1.4980 to 1.5000 | |
MFCD00008802 | |
09, IV, 715 | |
1.04 | |
Solubility in water: immiscible. Other solubilities: soluble in alcohols,esters,ethers and ketones | |
CC(C)(C)OOC(=O)C1=CC=CC=C1 | |
194.23 | |
6 mPa.s (20°C) | |
98% |
98% | |
8.0°C | |
1.0400g/mL | |
93°C | |
97.5% min. (GC) | |
C11H14O3 | |
C6H5CO2OC(CH3)3 | |
250 g | |
01,98; 02,54; 04,66; 07,49; 09,90; 13,58 | |
tert-butyl peroxybenzoate, tert-butyl perbenzoate, t-butyl perbenzoate, chaloxyd tbpb, perbutyl z, esperox 10, novox, trigonox c, tert-butyl peroxy benzoate, terc.butylperbenzoan | |
GJBRNHKUVLOCEB-UHFFFAOYSA-N | |
tert-butyl benzenecarboperoxoate | |
11966 | |
194.23 | |
Liquid |
Chemical Identifiers
614-45-9 | |
MFCD00008802 | |
tert-butyl peroxybenzoate, tert-butyl perbenzoate, t-butyl perbenzoate, chaloxyd tbpb, perbutyl z, esperox 10, novox, trigonox c, tert-butyl peroxy benzoate, terc.butylperbenzoan | |
tert-butyl benzenecarboperoxoate |
194.23 | |
GJBRNHKUVLOCEB-UHFFFAOYSA-N | |
11966 | |
CC(C)(C)OOC(=O)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Harmful if inhaled.
Heating may cause a fire.
May cause an allergic skin reaction.
Very toxic to aquatic life.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Keep/Store away from cl
GHS Signal Word: Danger
EINECSNumber : 210-382-2
RTECSNumber : SD9450000
TSCA : TSCA
RUO – Research Use Only