Please login to your online account to display your discounted pricing

tert-Butyl nitrite, 90%, pure, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  216225000

 View more versions of this product

Catalog No. AC216225000

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material tert-Butyl nitrite
CAS 540-80-7
Health Hazard 1 Danger
Infrared Spectrum Authentic
Purity Grade Pure
Physical Form Liquid
Assay ≥88% (GC)
Assay Percent Range 90%
Molecular Formula C4H9NO2
Linear Formula (CH3)3CNO2
Packaging Glass bottle
Formula Weight 103.12
Refractive Index 1.3670 to 1.3700 (20°C, 589nm)
Density 0.8600g/mL
Color Clear yellow
Quantity 500g

Specifications

Chemical Name or Material tert-Butyl nitrite
CAS 540-80-7
Purity Grade Pure
Assay ≥88% (GC)
Assay Percent Range 90%
Molecular Formula C4H9NO2
Linear Formula (CH3)3CNO2
Formula Weight 103.12
Accessories

Danger

SDS & Safety Documents