Please login to your online account to display your discounted pricing

tert-Butyl N-(4-piperidinylmethyl)carbamate, 97%

Chemical Identifiers

Chemical Name or Material tert-Butyl N-(4-piperidinylmethyl)carbamate
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H22N2O2
Formula Weight 214.31
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SPB08455CB

  SDS
fisher scientific company
SPB08455CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 214.31
Quantity 250mg
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H22N2O2
Chemical Name or Material tert-Butyl N-(4-piperidinylmethyl)carbamate
CAS 135632-53-0