Please login to your online account to display your discounted pricing

tert-Butyl N-(4-piperidinylmethyl)carbamate, 97%


Quantity 250mg

Manufacturer: fisher scientific company  SPB08455CB

 View more versions of this product

Catalog No. SPB08455CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 214.31
Quantity 250mg
Assay 95%
Assay Percent Range 95%
Chemical Name or Material tert-Butyl N-(4-piperidinylmethyl)carbamate
Molecular Formula C11H22N2O2
CAS 135632-53-0

Specifications

Chemical Name or Material tert-Butyl N-(4-piperidinylmethyl)carbamate
Assay 95%
Assay Percent Range 95%
Molecular Formula C11H22N2O2
Formula Weight 214.31