missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl N-(3-amino-3-thioxopropyl)carbamate, 97%, Maybridge

$621.89 - $621.89

Chemical Identifiers

CAS 77152-97-7
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C8H16N2O2S
Linear Formula (CH3)3COC(O)NH(CH2)2C(S)NH2
Formula Weight 204.29
MDL Number MFCD02180883
Chemical Name or Material Tert-butyl N-(3-amino-3-thioxopropyl)carbamate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
SPB08402DE Fisher Scientific Company
SPB08402DE
Amber Glass Bottle 5g Each for $621.89
Specifications Description & Specifications

Chemical Identifiers

77152-97-7
97%
(CH3)3COC(O)NH(CH2)2C(S)NH2
MFCD02180883
97%
97.0
C8H16N2O2S
204.29
Tert-butyl N-(3-amino-3-thioxopropyl)carbamate

Specifications

204.29
97.0
Amber Glass Bottle
97%
77152-97-7
97%
5g
C8H16N2O2S
MFCD02180883
(CH3)3COC(O)NH(CH2)2C(S)NH2
Tert-butyl N-(3-amino-3-thioxopropyl)carbamate