Please login to your online account to display your discounted pricing

tert-Butyl N-(3-amino-3-thioxopropyl)carbamate, 97%, Maybridge


Quantity 5g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SPB08402DE

 View more versions of this product

Catalog No. SPB08402DE

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 204.29
Quantity 5g
Assay 97%
CAS Min % 97.0
Chemical Name or Material Tert-butyl N-(3-amino-3-thioxopropyl)carbamate
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C8H16N2O2S
Linear Formula (CH3)3COC(O)NH(CH2)2C(S)NH2
MDL Number MFCD02180883
CAS 77152-97-7

Specifications

Chemical Name or Material Tert-butyl N-(3-amino-3-thioxopropyl)carbamate
CAS 77152-97-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H16N2O2S
Linear Formula (CH3)3COC(O)NH(CH2)2C(S)NH2
Formula Weight 204.29
MDL Number MFCD02180883