missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl N-(2-thienyl)carbamate, 97%, Maybridge

Chemical Identifiers

CAS 56267-50-6
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C9H13NO2S
Formula Weight 199.27
Chemical Name or Material Tert-butyl N-(2-thienyl)carbamate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC00045DA SDS Fisher Scientific Company
CC00045DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

56267-50-6
100.0
C9H13NO2S
Tert-butyl N-(2-thienyl)carbamate
96.5
97%
199.27
97%

Specifications

199.27
100.0
56267-50-6
96.5
Tert-butyl N-(2-thienyl)carbamate
1g
C9H13NO2S
Amber Glass Bottle
97%
97%