missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl N-(2-thienyl)carbamate, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC00045DA

 View more versions of this product

Catalog No. CC00045DA

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

199.27
97%
C9H13NO2S
Amber Glass Bottle
97%
1g
100.0
56267-50-6
96.5
Tert-butyl N-(2-thienyl)carbamate

Chemical Identifiers

56267-50-6
100.0
C9H13NO2S
Tert-butyl N-(2-thienyl)carbamate
96.5
97%
199.27
97%