missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
tert-Butyl N-(2-Hydroxypropyl)carbamate 98.0+%, TCI America™
$177.51 - $580.65
Chemical Identifiers
CAS | 95656-86-3 |
---|---|
Molecular Formula | C8H17NO3 |
Molecular Weight (g/mol) | 175.23 |
MDL Number | MFCD02093569 |
InChI Key | YNJCFDAODGKHAV-UHFFFAOYNA-N |
Synonym | 1-(Boc-amino)-2-propanol, 1-(tert-Butoxycarbonylamino)-2-propanol, N-(2-Hydroxypropyl)carbamic Acid tert-Butyl Ester |
PubChem CID | 9920508 |
IUPAC Name | tert-butyl N-(2-hydroxypropyl)carbamate |
SMILES | CC(O)CNC(=O)OC(C)(C)C |
Chemical Identifiers
95656-86-3 | |
175.23 | |
YNJCFDAODGKHAV-UHFFFAOYNA-N | |
9920508 | |
CC(O)CNC(=O)OC(C)(C)C |
C8H17NO3 | |
MFCD02093569 | |
1-(Boc-amino)-2-propanol, 1-(tert-Butoxycarbonylamino)-2-propanol, N-(2-Hydroxypropyl)carbamic Acid tert-Butyl Ester | |
tert-butyl N-(2-hydroxypropyl)carbamate |
Specifications
95656-86-3 | |
C8H17NO3 | |
1-(Boc-amino)-2-propanol, 1-(tert-Butoxycarbonylamino)-2-propanol, N-(2-Hydroxypropyl)carbamic Acid tert-Butyl Ester | |
CC(O)CNC(=O)OC(C)(C)C | |
175.23 | |
175.23 | |
5 g | |
tert-Butyl N-(2-Hydroxypropyl)carbamate |
Yellow | |
MFCD02093569 | |
YNJCFDAODGKHAV-UHFFFAOYNA-N | |
tert-butyl N-(2-hydroxypropyl)carbamate | |
9920508 | |
≥98.0% (N) | |
Liquid |
Safety and Handling
TSCA : No