Please login to your online account to display your discounted pricing

Tert-butyl monoperoxymaleate, ACROS Organics™


Quantity 1g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC378570010

 View more versions of this product

Catalog No. AC378570010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Viscous Paste
Formula Weight 188.18
Quantity 1g
Color White
Assay 23.0 to 27.0%
Chemical Name or Material tert-Butyl monoperoxymaleate
Molecular Formula C8H12O5
CAS 1931-62-0
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Heating may cause a fire.
Causes serious eye damage.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
CAS 105-76-0
Packaging Plastic bottle
MDL Number MFCD00082360
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material tert-Butyl monoperoxymaleate
CAS 1931-62-0
CAS 105-76-0
Assay 23.0 to 27.0%
Molecular Formula C8H12O5
Formula Weight 188.18
MDL Number MFCD00082360
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Heating may cause a fire.
Causes serious eye damage.
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.

SDS & Safety Documents