Please login to your online account to display your discounted pricing

tert-Butyl methyl sulfide, 97%, ACROS Organics™


Quantity 5mL

Manufacturer: thermo fisher scientific  337400050

 View more versions of this product

Catalog No. AC337400050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Liquid
Formula Weight 104.2
Quantity 5mL
Color Clear colorless
Chemical Name or Material tert-Butyl methyl sulfide
Infrared Spectrum Authentic
Name Note 97%
Assay Percent Range 97%
Molecular Formula C5H12S
Linear Formula (CH3)3CSCH3
Packaging Glass bottle
MDL Number MFCD00008840
CAS 6163-64-0
Refractive Index 1.4380 to 1.4410 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material tert-Butyl methyl sulfide
Name Note 97%
CAS 6163-64-0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H12S
Linear Formula (CH3)3CSCH3
Formula Weight 104.2
MDL Number MFCD00008840
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents