Please login to your online account to display your discounted pricing

tert-Butyl methyl sulfide, 97%, ACROS Organics™

$53.21 - $53.21

Chemical Identifiers

Chemical Name or Material tert-Butyl methyl sulfide
Name Note 97%
CAS 6163-64-0
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C5H12S
Linear Formula (CH3)3CSCH3
Formula Weight 104.20g/mol
MDL Number MFCD00008840
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC337400050

  SDS
thermo fisher scientific
337400050
5mL Each for $53.21
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4380 to 1.4410 (20°C, 589nm)
Color Clear colorless
Quantity 5mL
Chemical Name or Material tert-Butyl methyl sulfide
Name Note 97%
CAS 6163-64-0
Physical Form Clear colorless liquid
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C5H12S
Linear Formula (CH3)3CSCH3
Formula Weight 104.20g/mol
MDL Number MFCD00008840
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.