Please login to your online account to display your discounted pricing

tert-Butyl methyl ether, for HPLC, ACROS Organics™


Quantity 1L
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC389050010

 View more versions of this product

Catalog No. AC389050010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.8%
Formula Weight 88.15
Physical Form Liquid
Quantity 1L
Color Clear colorless
Density 0.7404g/mL
Purity Grade For HPLC
Merck Index 15, 6106
Acidity 0.0005 meq/g max.
Absorbance (1 cm cell vs HPLC−grade water)
Alkalinity 0.0002 meq/g max.
Linear Formula (CH3)3COCH3
Absorbance Conditions (1 cm cell vs HPLC-grade water)
Chemical Name or Material tert-Butyl methyl ether
Residue after Evaporation 0.0003% max.
Health Hazard 3 GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C5H12O
Health Hazard 2 GHS P Statement
Causes skin irritation.
Highly flammable liquid and vapor.
Packaging Glass bottle
Synonym 2-Methyl-2-methoxy propane
Water 0.05% max.
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00008812
CAS 1634-04-4

Specifications

Chemical Name or Material tert-Butyl methyl ether
CAS 1634-04-4
Purity Grade For HPLC
Assay ≥99.8%
Molecular Formula C5H12O
Linear Formula (CH3)3COCH3
Formula Weight 88.15
MDL Number MFCD00008812
Synonym 2-Methyl-2-methoxy propane
Accessories

GHS Signal Word: Danger

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes skin irritation.
Highly flammable liquid and vapor.

SDS & Safety Documents