Please login to your online account to display your discounted pricing

tert-Butyl methyl ether, For Spectroscopy Uvasol™, EMD Millipore

For spectroscopy Uvasol™

$363.74 - $363.74

Chemical Identifiers

Chemical Name or Material tert-Butyl methyl ether
CAS 1634-04-4
Molecular Formula (CH3)3COCH3
Formula Weight 88.15
UN Number 2398
Synonym MTBE, Methyl tert-butyl ether, 2-Methoxy-2-methylpropane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1019841000

 
emd millipore corporation
1019841000
Glass Bottle 1L Each for $363.74
Description & Specifications

Specifications

Boiling Point 55.3°C
Melting Point −108.6°C
Formula Weight 88.15
Quantity 1L
Solubility Information Soluble in water: 42g/L at 20°C
Color Colorless
Physical Form Liquid
Flash Point −28°C (−18.4°F)
Ignition Point 460°C (860°F)
UN Number 2398
Storage Information Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition. Protected from light.
Packaging Glass Bottle
Description The Uvasol™ solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The Uvasol™ solvents offer best UV transmittance.
Health Hazard 2 H315 Causes skin irritation.
Density 0.74g/cm3 at 20°C
Vapor Pressure 268 hPa (20°C)
Molecular Formula (CH3)3COCH3
Health Hazard 1 H225: Highly flammable liquid and vapour.
Chemical Name or Material tert-Butyl methyl ether
Odor Charactristic
Synonym MTBE, Methyl tert-butyl ether, 2-Methoxy-2-methylpropane
CAS 1634-04-4
Safety and Handling

H225: Highly flammable liquid and vapour.

H315 Causes skin irritation.