missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl methyl ether, 99%, pure, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC177040250

 View more versions of this product

Catalog No. AC177040250



Specifications

Specifications

tert-Butyl methyl ether
1634-04-4
C5H12O
MFCD00008812
04,333
2-methyl-2-methoxypropane
BZLVMXJERCGZMT-UHFFFAOYSA-N
2-methoxy-2-methylpropane
15413
88.15
99%
0.7404g/mL
Metal Drum
Solubility in water: 51g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5g/100g
0.01% Max. (Coulometric)
0.36 mPa.s (20°C)
White to Yellow
25L
99%
99%
(CH3)3COCH3
01, 381
15, 6106
Solubility in water: 51g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5g/100g
CC(C)(C)OC
88.15
CHEBI:27642
Powder or Needles
Pure
Authentic
1.3675 to 1.3695
0.7404
−28°C
54.0°C to 56.0°C
-110.0°C

Chemical Identifiers

1634-04-4
88.15
BZLVMXJERCGZMT-UHFFFAOYSA-N
15413
2-methoxy-2-methylpropane
C5H12O
MFCD00008812
2-methyl-2-methoxypropane
CHEBI:27642
CC(C)(C)OC
Safety and Handling

Safety and Handling

GHS P Statement
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents