missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
tert-Butyl methyl ether, 99+%, Extra Dry, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 375210010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
tert-Butyl methyl ether | |
Colorless | |
100 | |
54.0°C to 56.0°C | |
−28°C | |
AcroSeal™ Glass Bottle | |
1.3675 to 1.3695 | |
99% min. (GC) | |
C5H12O | |
MFCD00008812 | |
0.005% Max. | |
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
BZLVMXJERCGZMT-UHFFFAOYSA-N | |
2-methoxy-2-methylpropane | |
15413 | |
88.15 | |
Extra Dry |
-110.0°C | |
99 | |
0.7404g/mL | |
1 L | |
Authentic | |
Liquid | |
0.0005 % max. | |
0.7404 | |
(CH3)3COCH3 | |
0.36 mPa.s (20°C) | |
156106 | |
Solubility in water: 51g/L (20°C). Other solubilities: miscible with most organic solvents, unstable in acid solutions, soly of water in methyl-t-butyl ether : 1,5g/100g | |
CC(C)(C)OC | |
88.15 | |
CHEBI:27642 | |
99+% |
Chemical Identifiers
C5H12O | |
MFCD00008812 | |
tert-butyl methyl ether, methyl tert-butyl ether, mtbe, methyl t-butyl ether, t-butyl methyl ether, methyl tertiary-butyl ether, propane, 2-methoxy-2-methyl, methyl-tert-butyl ether, methyl-t-butyl ether, 2-methyl-2-methoxypropane | |
CHEBI:27642 | |
CC(C)(C)OC |
88.15 | |
BZLVMXJERCGZMT-UHFFFAOYSA-N | |
15413 | |
2-methoxy-2-methylpropane |
Safety and Handling
GHS H Statement
Causes skin irritation.
Highly flammable liquid and vapor.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Ground/Bond container and receiving equipment.
GHS Signal Word: Danger
EINECSNumber : 216-653-1
RUO – Research Use Only