missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl Methyl Ether

$47.74 - $47.74

Chemical Identifiers

CAS 1634-04-4
Chemical Name or Material tert-Butyl Methyl Ether
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
US-NV-2501 Ultra Scientific
NV2501
Ampule 1x1mL Each for $47.74
Specifications Description & Specifications

Chemical Identifiers

1634-04-4
tert-Butyl Methyl Ether

Specifications

1634-04-4
Butyl Methyl Ether
tert-Butyl Methyl Ether
Ampule
1x1mL