missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl Isocyanide 95.0+%, TCI America™
Supplier: TCI America B127425ML
Specifications
| tert-Butyl Isocyanide | |
| Colorless | |
| C5H9N | |
| 25 mL | |
| tert-butyl isocyanide, tert-butylisocyanide, t-butylisonitrile, tert-butylisonitrile, tbin, propane, 2-isocyano-2-methyl, 2-methyl-2-isocyanopropane, 2-methylpropyl isocyanide, t-butyl isocyanide, 2-isocyano-2-methyl-propane | |
| CC(C)(C)[N+]#[C-] | |
| 83.134 | |
| 83.13 | |
| Liquid |
| 7188-38-7 | |
| 92°C | |
| MFCD00000002 | |
| 1992 | |
| FAGLEPBREOXSAC-UHFFFAOYSA-N | |
| 2-isocyano-2-methylpropane | |
| 23577 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 7188-38-7 | |
| 83.134 | |
| FAGLEPBREOXSAC-UHFFFAOYSA-N | |
| 23577 | |
| CC(C)(C)[N+]#[C-] |
| C5H9N | |
| MFCD00000002 | |
| tert-butyl isocyanide, tert-butylisocyanide, t-butylisonitrile, tert-butylisonitrile, tbin, propane, 2-isocyano-2-methyl, 2-methyl-2-isocyanopropane, 2-methylpropyl isocyanide, t-butyl isocyanide, 2-isocyano-2-methyl-propane | |
| 2-isocyano-2-methylpropane |
Safety and Handling
RTECSNumber : EQ7102500
TSCA : No