missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl isocyanate, 97%, ACROS Organics™

Chemical Identifiers

CAS 1609-86-5
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
MDL Number MFCD00002035
InChI Key MGOLNIXAPIAKFM-UHFFFAOYSA-N
Synonym tert-butyl isocyanate, tert-butylisocyanate, propane, 2-isocyanato-2-methyl, t-butyl isocyanate, t-butylisocyanate, unii-2rhq1bs36d, 2-isocyanato-2-methyl-propane, isocyanic acid, tert-butyl ester, 2rhq1bs36d, 1,1-dimethylethyl isocyanate
PubChem CID 62412
IUPAC Name 2-isocyanato-2-methylpropane
SMILES CC(C)(C)N=C=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC162961000 SDS View Documents Acros Organics
AC162961000
100g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1609-86-5
99.133
MGOLNIXAPIAKFM-UHFFFAOYSA-N
62412
CC(C)(C)N=C=O
C5H9NO
MFCD00002035
tert-butyl isocyanate, tert-butylisocyanate, propane, 2-isocyanato-2-methyl, t-butyl isocyanate, t-butylisocyanate, unii-2rhq1bs36d, 2-isocyanato-2-methyl-propane, isocyanic acid, tert-butyl ester, 2rhq1bs36d, 1,1-dimethylethyl isocyanate
2-isocyanato-2-methylpropane

Specifications

0.8440g/mL
Authentic
1.3850 to 1.3870 (20°C, 589nm)
Colorless
1609-86-5
C5H9NO
MFCD00002035
MGOLNIXAPIAKFM-UHFFFAOYSA-N
2-isocyanato-2-methylpropane
62412
Liquid
tert-Butyl isocyanate
−4°C
Glass bottle
84.0°C
100g
97%
(CH3)3CNCO
tert-butyl isocyanate, tert-butylisocyanate, propane, 2-isocyanato-2-methyl, t-butyl isocyanate, t-butylisocyanate, unii-2rhq1bs36d, 2-isocyanato-2-methyl-propane, isocyanic acid, tert-butyl ester, 2rhq1bs36d, 1,1-dimethylethyl isocyanate
CC(C)(C)N=C=O
99.133
99.13
97%
Safety and Handling

Safety and Handling

GHS H Statement
Harmful in contact with skin. Causes severe skin burns and eye damage. Fatal if inhaled. Harmful if swallowed. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Highly flammable liquid and vapor. Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Danger