Please login to your online account to display your discounted pricing

Alfa Aesar™ tert-Butyl isocyanate, 97%

$34.07 - $102.23

Chemical Identifiers

Chemical Name or Material tert-Butyl isocyanate
CAS 1609-86-5
Molecular Formula (CH3)3CNCO
Formula Weight 99.13g/mol
MDL Number MFCD00002035
UN Number UN2484
Beilstein 969479
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1337106

 
alfa aesar™
L13371-06
5g Each for $34.07

AAL1337114

 
alfa aesar™
L13371-14
25g Each for $102.23

AAL1337122

 
alfa aesar™
L13371-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
Boiling Point 85° to 86°C
Flash Point −4°C (24°F)
Odor Acrid
Quantity 5g
Chemical Name or Material tert-Butyl isocyanate
CAS 1609-86-5
Assay 97%
Molecular Formula (CH3)3CNCO
Formula Weight 99.13g/mol
MDL Number MFCD00002035
UN Number UN2484
Beilstein 969479
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H301-H330-H334-H315-H319-H335
Highly flammable liquid and vapor.
Toxic if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TERT-BUTYL ISOCYANATE
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H301-H330-H334-H315-H319-H335
Highly flammable liquid and vapor.
Toxic if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C