Learn More
tert-Butyl hydrogen malonate, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2622306
Description
tert-Butyl hydrogen malonate may be used in the preparation of dendritic precursor to asymmetric methanofullerenes, hapten-3,6-(O,S-dimethylthiophosphoramido)-6-oxohexanoic acid, hapten-4,3-(O,S-dimethylthiophosphoramido)-3-oxopropanoic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstert-Butyl hydrogen malonate may be used in the preparation of dendritic precursor to asymmetric methanofullerenes, hapten-3,6-(O,S-dimethylthiophosphoramido)-6-oxohexanoic acid, hapten-4,3-(O,S-dimethylthiophosphoramido)-3-oxopropanoic acid.
Solubility
Soluble in ethanol.
Notes
Store in a cool, dry condition in a well sealed container.
Specifications
tert-Butyl hydrogen malonate | |
19°C to 20°C | |
90°C (2 mmHg) | |
C7H12O4 | |
MFCD00191886 | |
1765457 | |
Soluble in ethanol. | |
CC(C)(C)OC(=O)CC(=O)O | |
160.169 | |
160.17 |
40052-13-9 | |
1.04 | |
91°C (195°F) | |
1.426 | |
5 g | |
3-tert-butoxy-3-oxopropanoic acid, malonic acid mono-tert-butyl ester, mono-tert-butyl malonate, 2-tert-butoxycarbonyl acetic acid, tert-butyl hydrogen malonate, mono-t-butyl malonate, pubchem13754, malonicacidmono-tert-butylester, malonic acid mono t-butyl ester | |
NGGGZUAEOKRHMA-UHFFFAOYSA-N | |
3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid | |
545853 | |
96% |
Chemical Identifiers
40052-13-9 | |
160.169 | |
NGGGZUAEOKRHMA-UHFFFAOYSA-N | |
545853 | |
CC(C)(C)OC(=O)CC(=O)O |
C7H12O4 | |
MFCD00191886 | |
3-tert-butoxy-3-oxopropanoic acid, malonic acid mono-tert-butyl ester, mono-tert-butyl malonate, 2-tert-butoxycarbonyl acetic acid, tert-butyl hydrogen malonate, mono-t-butyl malonate, pubchem13754, malonicacidmono-tert-butylester, malonic acid mono t-butyl ester | |
3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H330-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only