missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl Ethyl Ether, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC403531000

 View more versions of this product

Catalog No. AC403531000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

tert-Butyl ethyleether
0.75g/mL
Glass bottle
Clear colorless
≥96.0%
97%
(CH3)3COC2H5
MFCD00009225
637-92-3
Authentic
1.3746 to 1.3766 (20°C, 589nm)
Liquid
100g
C6H14O
102.18
15, 3829

Chemical Identifiers

97%
(CH3)3COC2H5
MFCD00009225
≥96.0%
C6H14O
102.18
tert-Butyl ethyleether

Safety and Handling

SDS & Safety Documents

GHS P Statement
Highly flammable liquid and vapor.
May cause drowsiness or dizziness.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger