Please login to your online account to display your discounted pricing

tert-Butyl ethyl malonate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material tert-Butyl ethyl malonate
Assay Percent Range 97%
Molecular Formula C9H16O4
Linear Formula (CH3)3COCCHCOCH5
Formula Weight 188.22
MDL Number MFCD00009193
Synonym Malonic acid tert-butyl ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC339400250

  SDS
thermo fisher scientific
AC339400250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 188.22
Quantity 25g
Density 1.001g/mL
Assay Percent Range 97%
Linear Formula (CH3)3COCCHCOCH5
CAS 32864-38-3
Chemical Name or Material tert-Butyl ethyl malonate
Packaging Glass bottle
Molecular Formula C9H16O4
MDL Number MFCD00009193
Synonym Malonic acid tert-butyl ethyl ester