missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ tert-Butyl ethyl malonate, 95%

Manufacturer:  Alfa Aesar™ A1192722

 View more versions of this product

Catalog No. AAA1192722


 

Description

Description

Applications
tert-Butyl ethyl malonate is used in the preparation of polar ester-functionalized aliphatic polysulfone. It is also a useful reagent for organic synthesis.

Solubility
Miscible with most common solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

tert-Butyl ethyl malonate
95%
MFCD00009193
Malonic acid tert-butyl ethyl ester
CCOC(=O)CC(=O)OC(C)(C)C
188.223
188.22
87°C (188°F)
32864-38-3
C9H16O4
1776681
OCOBFMZGRJOSOU-UHFFFAOYSA-N
3-O-tert-butyl 1-O-ethyl propanedioate
96345
98°C to 100°C (22mmHg)
100g

Chemical Identifiers

32864-38-3
188.223
OCOBFMZGRJOSOU-UHFFFAOYSA-N
96345
C9H16O4
MFCD00009193
Malonic acid tert-butyl ethyl ester
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents