missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ tert-Butyl cyanoacetate, 98%

$56.59 - $624.55

Chemical Identifiers

CAS 1116-98-9
Molecular Formula C7H11NO2
Molecular Weight (g/mol) 141.17
MDL Number MFCD00001938
InChI Key BFNYNEMRWHFIMR-UHFFFAOYSA-N
Synonym tert-butyl cyanoacetate, t-butyl cyanoacetate, acetic acid, cyano-, 1,1-dimethylethyl ester, cyanoacetic acid tert-butyl ester, acetic acid, cyano-, tert-butyl ester, acetic acid, 2-cyano-, 1,1-dimethylethyl ester, tert-butylcyanacetat, t-butylcyanoacetate, tert-butylcyanoacetate, tert butyl cyanoacetate
PubChem CID 70693
IUPAC Name tert-butyl 2-cyanoacetate
SMILES CC(C)(C)OC(=O)CC#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0012306 View Documents Alfa Aesar™
L0012306
5g Each for $56.59
 
AAL0012314 View Documents Alfa Aesar™
L0012314
25g Each for $181.97
 
AAL0012322 View Documents Alfa Aesar™
L0012322
100g Each for $624.55
 
Description

Description

Applications
tert-Butyl cyanoacetate is used in the synthesis of vinylogous urea. It is also used as a new additive for the sugar nucleoside base coupling step en route to DAPD with improved β-selectivity.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

1116-98-9
141.17
BFNYNEMRWHFIMR-UHFFFAOYSA-N
70693
CC(C)(C)OC(=O)CC#N
C7H11NO2
MFCD00001938
tert-butyl cyanoacetate, t-butyl cyanoacetate, acetic acid, cyano-, 1,1-dimethylethyl ester, cyanoacetic acid tert-butyl ester, acetic acid, cyano-, tert-butyl ester, acetic acid, 2-cyano-, 1,1-dimethylethyl ester, tert-butylcyanacetat, t-butylcyanoacetate, tert-butylcyanoacetate, tert butyl cyanoacetate
tert-butyl 2-cyanoacetate

Specifications

91°C (195°F)
67°C to 68°C (1.5mmHg)
1116-98-9
MFCD00001938
tert-butyl cyanoacetate, t-butyl cyanoacetate, acetic acid, cyano-, 1,1-dimethylethyl ester, cyanoacetic acid tert-butyl ester, acetic acid, cyano-, tert-butyl ester, acetic acid, 2-cyano-, 1,1-dimethylethyl ester, tert-butylcyanacetat, t-butylcyanoacetate, tert-butylcyanoacetate, tert butyl cyanoacetate
CC(C)(C)OC(=O)CC#N
141.17
141.17
tert-Butyl cyanoacetate
Odorless
5g
C7H11NO2
1755933
BFNYNEMRWHFIMR-UHFFFAOYSA-N
tert-butyl 2-cyanoacetate
70693
98%
Safety and Handling

Safety and Handling

GHS H Statement
H301-H227
Toxic if swallowed.
Combustible liquid.

GHS P Statement
P210-P280-P301+P310-P405-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA