Learn More
tert-Butyl cyanoacetate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0012306
Description
tert-Butyl cyanoacetate is used in the synthesis of vinylogous urea. It is also used as a new additive for the sugar nucleoside base coupling step en route to DAPD with improved β-selectivity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstert-Butyl cyanoacetate is used in the synthesis of vinylogous urea. It is also used as a new additive for the sugar nucleoside base coupling step en route to DAPD with improved β-selectivity.
Solubility
Difficult to mix.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
tert-Butyl cyanoacetate | |
0.987 | |
91°C (195°F) | |
C7H11NO2 | |
MFCD00001938 | |
1755933 | |
Difficult to mix. | |
CC(C)(C)OC(=O)CC#N | |
141.17 | |
141.17 |
1116-98-9 | |
67°C to 68°C (1.5 mmHg) | |
Odorless | |
1.42 | |
5 g | |
tert-butyl cyanoacetate, t-butyl cyanoacetate, acetic acid, cyano-, 1,1-dimethylethyl ester, cyanoacetic acid tert-butyl ester, acetic acid, cyano-, tert-butyl ester, acetic acid, 2-cyano-, 1,1-dimethylethyl ester, tert-butylcyanacetat, t-butylcyanoacetate, tert-butylcyanoacetate, tert butyl cyanoacetate | |
BFNYNEMRWHFIMR-UHFFFAOYSA-N | |
tert-butyl 2-cyanoacetate | |
70693 | |
98% |
Chemical Identifiers
1116-98-9 | |
141.17 | |
BFNYNEMRWHFIMR-UHFFFAOYSA-N | |
70693 | |
CC(C)(C)OC(=O)CC#N |
C7H11NO2 | |
MFCD00001938 | |
tert-butyl cyanoacetate, t-butyl cyanoacetate, acetic acid, cyano-, 1,1-dimethylethyl ester, cyanoacetic acid tert-butyl ester, acetic acid, cyano-, tert-butyl ester, acetic acid, 2-cyano-, 1,1-dimethylethyl ester, tert-butylcyanacetat, t-butylcyanoacetate, tert-butylcyanoacetate, tert butyl cyanoacetate | |
tert-butyl 2-cyanoacetate |
Safety and Handling
GHS H Statement
H301-H227
Toxic if swallowed.
Combustible liquid.
P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
H227-H302
EINECSNumber : 214-243-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only