Learn More
tert-Butyl chloroacetate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 347230250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
tert-Butyl chloroacetate | |
1.0530g/mL | |
46°C | |
96% min. (GC) | |
C6H11ClO2 | |
ClCH2CO2C(CH3)3 | |
25g | |
01,86; 03,36 | |
15, 1564 | |
Solubility in water: <0.1g/L (20°C). Other solubilities: soluble in isopropanol,1,2-dichloroethane,toluene | |
CC(C)(C)OC(=O)CCl | |
150 | |
150 |
107-59-5 | |
48.0°C to 49.0°C (11.0mmHg) | |
Authentic | |
Glass bottle | |
1.4220 to 1.4240 | |
MFCD00000930 | |
02, III, 444 | |
1.053 | |
tert-butyl chloroacetate, chloroacetic acid tert-butyl ester, acetic acid, chloro-, 1,1-dimethylethyl ester, t-butyl chloroacetate, tert-butylchloroacetate, acetic acid, chloro-, tert-butyl ester, chloroacetic acid t-butyl ester, acetic acid, 2-chloro-, 1,1-dimethylethyl ester, t-butylchloroacetate, t-butyl-chloroacetate | |
KUYMVWXKHQSIAS-UHFFFAOYSA-N | |
tert-butyl 2-chloroacetate | |
66052 | |
97% |
Chemical Identifiers
107-59-5 | |
150 | |
KUYMVWXKHQSIAS-UHFFFAOYSA-N | |
66052 | |
CC(C)(C)OC(=O)CCl |
C6H11ClO2 | |
MFCD00000930 | |
tert-butyl chloroacetate, chloroacetic acid tert-butyl ester, acetic acid, chloro-, 1,1-dimethylethyl ester, t-butyl chloroacetate, tert-butylchloroacetate, acetic acid, chloro-, tert-butyl ester, chloroacetic acid t-butyl ester, acetic acid, 2-chloro-, 1,1-dimethylethyl ester, t-butylchloroacetate, t-butyl-chloroacetate | |
tert-butyl 2-chloroacetate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Very toxic to aquatic life with long lasting effects.
Harmful in contact with skin.
Harmful if swallowed.
Toxic if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
EINECSNumber : 203-506-1
RUO â Research Use Only