missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl carbazate, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 107880250

 View more versions of this product

Catalog No. AC107880250


 

Specifications Description & Specifications

Specifications

Crystalline Lumps
≥98.5%
White to slightly yellow
01,85; 02,46; 11,88
Authentic
C5H12N2O2
99%
H2NNHCO2C(CH3)3
99%
132.16
25g
37° to 41°C
BOC-hydrazide
Glass bottle
870-46-2
tert-Butyl carbazate
MFCD00007593

Chemical Identifiers

870-46-2
C5H12N2O2
132.16
BOC-hydrazide
≥98.5%
99%
H2NNHCO2C(CH3)3
MFCD00007593
tert-Butyl carbazate, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
Toxic in contact with skin.
Flammable solid.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 212-795-3

TSCA : TSCA