Learn More
tert-Butyl bromoacetate, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 154342500
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
tert-Butyl bromoacetate | |
5292-43-3 | |
50°C (10.0mmHg) | |
Authentic | |
Glass bottle | |
1.4440 to 1.446 | |
MFCD00000188 | |
02, I, 96 | |
tert-butyl bromoacetate, t-butyl bromoacetate, acetic acid, bromo-, 1,1-dimethylethyl ester, bromoacetic acid tert-butyl ester, tert-butylbromoacetate, acetic acid, bromo-, tert-butyl ester, brch2c o oc ch3 3, acetic acid, 2-bromo-, 1,1-dimethylethyl ester, bromo-acetic acid tert-butyl ester, tert-butylbromacetat | |
BNWCETAHAJSBFG-UHFFFAOYSA-N | |
tert-butyl 2-bromoacetate | |
79177 | |
99% |
99% | |
1.3380g/mL | |
49°C | |
98.5% min. (GC) | |
C6H11BrO2 | |
BrCH2CO2C(CH3)3 | |
250mL | |
1.338 | |
Solubility in water: immiscible | |
CC(C)(C)OC(=O)CBr | |
195.06 | |
195.06 |
Safety and Handling
GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
EINECSNumber : 226-133-6
Recommended Storage : Refrigerator +4°C
RUO â Research Use Only