Learn More
tert-Butyl bromoacetate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1491709
Description
tert-Butyl bromoacetate is used as an intermediate in the production of dyes, pharmaceuticals, agrochemicals and in organic compounds. It finds application as an alkylating agent. It plays an important role to prepare (2-oxo-4-vinyl-azetidin-1-yl)-acetic acid tert-butyl ester and in collagenase inhibitors synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstert-Butyl bromoacetate is used as an intermediate in the production of dyes, pharmaceuticals, agrochemicals and in organic compounds. It finds application as an alkylating agent. It plays an important role to prepare (2-oxo-4-vinyl-azetidin-1-yl)-acetic acid tert-butyl ester and in collagenase inhibitors synthesis.
Solubility
Miscible with ethanol, chloroform and ethyl acetate. Immiscible with water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
Specifications
tert-Butyl bromoacetate | |
1.33 | |
49°C (120°F) | |
C6H11BrO2 | |
BrCH2CO2C(CH3)3 | |
10 g | |
1753010 | |
Miscible with ethanol,chloroform and ethyl acetate. Immiscible with water. | |
CC(C)(C)OC(=O)CBr | |
195.06 | |
195.06 |
5292-43-3 | |
175°C | |
Pungent | |
1.445 | |
MFCD00000188 | |
UN2924 | |
tert-butyl bromoacetate, t-butyl bromoacetate, acetic acid, bromo-, 1,1-dimethylethyl ester, bromoacetic acid tert-butyl ester, tert-butylbromoacetate, acetic acid, bromo-, tert-butyl ester, brch2c o oc ch3 3, acetic acid, 2-bromo-, 1,1-dimethylethyl ester, bromo-acetic acid tert-butyl ester, tert-butylbromacetat | |
BNWCETAHAJSBFG-UHFFFAOYSA-N | |
tert-butyl 2-bromoacetate | |
79177 | |
98% |
Chemical Identifiers
5292-43-3 | |
195.06 | |
BNWCETAHAJSBFG-UHFFFAOYSA-N | |
79177 | |
CC(C)(C)OC(=O)CBr |
C6H11BrO2 | |
MFCD00000188 | |
tert-butyl bromoacetate, t-butyl bromoacetate, acetic acid, bromo-, 1,1-dimethylethyl ester, bromoacetic acid tert-butyl ester, tert-butylbromoacetate, acetic acid, bromo-, tert-butyl ester, brch2c o oc ch3 3, acetic acid, 2-bromo-, 1,1-dimethylethyl ester, bromo-acetic acid tert-butyl ester, tert-butylbromacetat | |
tert-butyl 2-bromoacetate |
Safety and Handling
GHS H Statement
H314-H318-H226-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
EINECSNumber : 226-133-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only