missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl Benzoate 98.0+%, TCI America™

Manufacturer:  TCI America B0067500ML

 View more versions of this product

Catalog No. B0067500ML



Specifications

Specifications

tert-Butyl Benzoate
Yellow
774-65-2
MFCD00048244
LYDRKKWPKKEMNZ-UHFFFAOYSA-N
tert-butyl benzoate
69886
178.23
≥98.0% (GC)
54°C
500mL
C11H14O2
Benzoic Acid tert-Butyl Ester
CC(C)(C)OC(=O)C1=CC=CC=C1
178.231
CHEBI:34034
Clear Liquid at 20°C

Chemical Identifiers

774-65-2
178.231
LYDRKKWPKKEMNZ-UHFFFAOYSA-N
69886
tert-butyl benzoate
C11H14O2
MFCD00048244
Benzoic Acid tert-Butyl Ester
CHEBI:34034
CC(C)(C)OC(=O)C1=CC=CC=C1
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1356

TSCA : No

Documents