missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ tert-Butyl alcohol, 99%

Manufacturer:  Alfa Aesar™ L12338AK

 View more versions of this product

Catalog No. AAL12338AK



Description

Description

Applications
tert-Butyl alcohol is used as a solvent and ethanol denaturant. It is an active ingredient of paint remover. It acts as an intermediate in the production of methyl tertiary butyl ether and ethyl tertiary butyl ether by reaction with methanol and ethanol. It is also used in the preparation of tert-butyl chloride, tert-butyl hydroperoxide, isobutylene and methyl methacrylate. It serves as a fuel additive as well as a gasoline octane booster.

Solubility
Miscible with water, ethanol, ethyl ether and chloroform.

Notes
Incompatible with strong oxidizing agents, copper, alkali metals and aluminum.
Specifications

Specifications

tert-Butyl alcohol
11°C (52°F)
83°C
250mL
C4H10O
UN1120
14,1542
Miscible with water,ethanol,ethyl ether and chloroform.
CC(C)(C)O
74.123
CHEBI:45895
99%
0.786
1.387
25°C
75-65-0
MFCD00004464
906698
1,1-dimethylethanol
DKGAVHZHDRPRBM-UHFFFAOYSA-N
2-methylpropan-2-ol
6386
74.12

Chemical Identifiers

75-65-0
74.123
DKGAVHZHDRPRBM-UHFFFAOYSA-N
6386
2-methylpropan-2-ol
C4H10O
MFCD00004464
1,1-dimethylethanol
CHEBI:45895
CC(C)(C)O
Safety and Handling

Safety and Handling

GHS H Statement
H225-H332-H319-H335
Highly flammable liquid and vapor.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c

H225-H319-H332-H335-H336

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: BUTANOLS

EINECSNumber : 200-889-7

RTECSNumber : EO1925000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents