Please login to your online account to display your discounted pricing

tert-Butyl acrylate, 99%, stabilized, ACROS Organics™


Quantity 5L
Packaging Plastic Jerrycan

Manufacturer: thermo fisher scientific  AC371130051

 View more versions of this product

Catalog No. AC371130051

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 128.17
Quantity 5L
Color Clear
Packaging Plastic Jerrycan
Density 0.8830g/mL
Beilstein 02, IV, 1465
Chemical Name or Material tert-Butyl acrylate
Name Note 99%
Infrared Spectrum Authentic
Molecular Formula C7H12O2
Refractive Index 1.4100 to 1.4120 (20°C, 589nm)
Linear Formula H2C=CHCO2C(CH3)3
MDL Number MFCD00008809
CAS 1663-39-4
Stabilizer 10 to 20ppm MEHQ
Health Hazard 1 Danger
CAS 150-76-5
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 99%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Highly flammable liquid and vapor.

Specifications

Chemical Name or Material tert-Butyl acrylate
Name Note 99%
CAS 1663-39-4
CAS 150-76-5
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H12O2
Linear Formula H2C=CHCO2C(CH3)3
Formula Weight 128.17
MDL Number MFCD00008809
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Highly flammable liquid and vapor.

SDS & Safety Documents