missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl acrylate, 99%, stab. with 15ppm 4-methoxyphenol, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L17169AU

 View more versions of this product

Catalog No. AAL17169AU



Description

Description

tert-Butyl acrylate is a monomer used in the production of poly(tert-butyl acrylate) polymers, poly(acrylic acid) polyelectrolyte brushes and 3-(3-methyl-but-3-enyloxy)-propionic acid tert-butyl ester. It is also used in paints, coatings, high polymer paper conditioner, nursing polymer products and carbonyl reforming polyolefin. Further, it serves as an important raw material in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl acrylate is a monomer used in the production of poly(tert-butyl acrylate) polymers, poly(acrylic acid) polyelectrolyte brushes and 3-(3-methyl-but-3-enyloxy)-propionic acid tert-butyl ester. It is also used in paints, coatings, high polymer paper conditioner, nursing polymer products and carbonyl reforming polyolefin. Further, it serves as an important raw material in organic synthesis.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

tert-Butyl acrylate
1663-39-4
0.883
11°C (52°F)
1.411
MFCD00008809
UN3272
tert-butyl acrylate, t-butyl acrylate, tert-butyl propenoate, 2-propenoic acid, 1,1-dimethylethyl ester, acrylic acid tert-butyl ester, tert-butylacrylate, acrylic acid, tert-butyl ester, unii-92m2cd1o3g, ccris 7035, 2-propenoic acid tert-butyl ester
ISXSCDLOGDJUNJ-UHFFFAOYSA-N
tert-butyl prop-2-enoate
15458
99%
stabilized with 15ppm 4-methoxyphenol
∼-69°C
121°C
C7H12O2
CH2=CHCOOC(CH3)3
1000 mL
1742329
Slightly miscible with water.
CC(C)(C)OC(=O)C=C
128.17
128.17

Chemical Identifiers

1663-39-4
128.17
ISXSCDLOGDJUNJ-UHFFFAOYSA-N
15458
CC(C)(C)OC(=O)C=C
C7H12O2
MFCD00008809
tert-butyl acrylate, t-butyl acrylate, tert-butyl propenoate, 2-propenoic acid, 1,1-dimethylethyl ester, acrylic acid tert-butyl ester, tert-butylacrylate, acrylic acid, tert-butyl ester, unii-92m2cd1o3g, ccris 7035, 2-propenoic acid tert-butyl ester
tert-butyl prop-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H225-H302-H312-H332-H315-H317-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P333+P313-P363-P370+P378q-P501c

H225-H302+H312+H332-H315-H317-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 216-768-7

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only