missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ tert-Butyl acetoacetate, 98+%

Manufacturer:  Alfa Aesar™ A1111522

 View more versions of this product

Catalog No. AAA1111522



Description

Description

Applications
tert-Butyl acetoacetate is used as intermediate for the syntheses of different organic chemicals. Raw material in pharmaceutical Industry, In agrochemical industry. Reagent for Acyloin synthesis and ∞,β - unsaturated ketones synthesis.

Solubility
Soluble in water (9g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

tert-Butyl acetoacetate
Pleasant
-38°C
5.7
C8H14O3
1680303
JKUYRAMKJLMYLO-UHFFFAOYSA-N
tert-butyl 3-oxobutanoate
15538
≥98%
76°C (169°F)
71°C to 72°C (11mmHg)
100g
1694-31-1
MFCD00008811
tert-butyl acetoacetate, t-butyl acetoacetate, butanoic acid, 3-oxo-, 1,1-dimethylethyl ester, acetoacetic acid, tert-butyl ester, tert-butyl 3-oxobutyrate, acetoacetic acid tert-butyl ester, unii-1d3dos61gx, tbaa, 1d3dos61gx, t-butylacetoacetate
CC(=O)CC(=O)OC(C)(C)C
158.197
158.2

Chemical Identifiers

1694-31-1
158.197
JKUYRAMKJLMYLO-UHFFFAOYSA-N
15538
CC(=O)CC(=O)OC(C)(C)C
C8H14O3
MFCD00008811
tert-butyl acetoacetate, t-butyl acetoacetate, butanoic acid, 3-oxo-, 1,1-dimethylethyl ester, acetoacetic acid, tert-butyl ester, tert-butyl 3-oxobutyrate, acetoacetic acid tert-butyl ester, unii-1d3dos61gx, tbaa, 1d3dos61gx, t-butylacetoacetate
tert-butyl 3-oxobutanoate
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents