Learn More
tert-Butyl acetate, 99%, Thermo Scientific Chemicals
$410.72 - $410.72
Chemical Identifiers
CAS | 540-88-5 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00008807 |
InChI Key | WMOVHXAZOJBABW-UHFFFAOYSA-N |
Synonym | t-butyl acetate, acetic acid tert-butyl ester, tert-butyl ethanoate, acetic acid, 1,1-dimethylethyl ester, texaco lead appreciator, acetic acid, tert-butyl ester, tert-butylacetate, tert butyl acetate, 1,1-dimethylethyl acetate, 1,1-dimethyl acetate |
PubChem CID | 10908 |
IUPAC Name | tert-butyl acetate |
SMILES | CC(=O)OC(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC107765000
|
Thermo Scientific Chemicals
107765000 |
500 mL | Glass bottle |
Each for $410.72
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
540-88-5 | |
116.16 | |
WMOVHXAZOJBABW-UHFFFAOYSA-N | |
10908 | |
CC(=O)OC(C)(C)C |
C6H12O2 | |
MFCD00008807 | |
t-butyl acetate, acetic acid tert-butyl ester, tert-butyl ethanoate, acetic acid, 1,1-dimethylethyl ester, texaco lead appreciator, acetic acid, tert-butyl ester, tert-butylacetate, tert butyl acetate, 1,1-dimethylethyl acetate, 1,1-dimethyl acetate | |
tert-butyl acetate |
Specifications
540-88-5 | |
0.8600g/mL | |
15°C | |
98.5% min. (GC) | |
C6H12O2 | |
CH3CO2C(CH3)3 | |
500 mL | |
01,82 | |
15, 1539 | |
Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether | |
CC(=O)OC(C)(C)C | |
116.16 | |
116.16 | |
Liquid |
Colorless | |
98.0°C | |
Authentic | |
Glass bottle | |
1.3850 to 1.3870 | |
MFCD00008807 | |
02, 131 | |
0.86 | |
t-butyl acetate, acetic acid tert-butyl ester, tert-butyl ethanoate, acetic acid, 1,1-dimethylethyl ester, texaco lead appreciator, acetic acid, tert-butyl ester, tert-butylacetate, tert butyl acetate, 1,1-dimethylethyl acetate, 1,1-dimethyl acetate | |
WMOVHXAZOJBABW-UHFFFAOYSA-N | |
tert-butyl acetate | |
10908 | |
99% | |
tert-Butyl acetate, 99+% |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Harmful if inhaled.
May cause respiratory irritation.
May cause drowsiness or dizziness.
Repeated exposure may cause skin dryness or cracking.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Avoid breathing dust/fume/gas/mist/vapors/spray.
GHS Signal Word: Danger
EINECSNumber : 208-760-7
RTECSNumber : AF7400000
TSCA : TSCA
RUO – Research Use Only