Please login to your online account to display your discounted pricing

tert-Butyl 6-fluoro-1H-pyrazolo[3,4-b]pyridine-3-carboxylate, ≥97%, Maybridge

Chemical Identifiers

Chemical Name or Material tert-Butyl 6-fluoro-1H-pyrazolo[3, 4-b]pyridine-3-carboxylate
CAS 155601-71-1
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H12FN3O2
Formula Weight 237.23
MDL Number MFCD06797502
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO07504CB

  SDS
fisher scientific company
MO07504CB
Amber Glass Bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 237.23
Quantity 250mg
Assay 97%
CAS Min % 97.0
Chemical Name or Material tert-Butyl 6-fluoro-1H-pyrazolo[3, 4-b]pyridine-3-carboxylate
Molecular Formula C11H12FN3O2
MDL Number MFCD06797502
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 155601-71-1