missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl 4-formyl-3,5-dimethyl-1H-pyrazole-1-carboxylate, 97%, Maybridge

Manufacturer:  Fisher Scientific Company CC31904DE

 View more versions of this product

Catalog No. CC31904DE

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

224.26
97%
C11H16N2O3
Tert-butyl 4-formyl-3, 5-dimethyl-1H-pyrazole-1-carboxylate
844891-13-0
5g
100.0
97%
96.5
MFCD06659076

Chemical Identifiers

844891-13-0
100.0
C11H16N2O3
MFCD06659076
97%
96.5
97%
224.26
Tert-butyl 4-formyl-3, 5-dimethyl-1H-pyrazole-1-carboxylate

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.