missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

tert-Butyl 4-formyl-3,5-dimethyl-1H-pyrazole-1-carboxylate, 97%, Maybridge

Chemical Identifiers

CAS 844891-13-0
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C11H16N2O3
Formula Weight 224.26
MDL Number MFCD06659076
Chemical Name or Material Tert-butyl 4-formyl-3, 5-dimethyl-1H-pyrazole-1-carboxylate
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CC31904DE Fisher Scientific Company
CC31904DE
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC31904DA SDS Fisher Scientific Company
CC31904DA
Amber glass bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

844891-13-0
100.0
C11H16N2O3
MFCD06659076
97%
96.5
97%
224.26
Tert-butyl 4-formyl-3, 5-dimethyl-1H-pyrazole-1-carboxylate

Specifications

224.26
100.0
97%
844891-13-0
Tert-butyl 4-formyl-3, 5-dimethyl-1H-pyrazole-1-carboxylate
5g
C11H16N2O3
96.5
MFCD06659076
97%

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.